missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI1 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803805
This item is not returnable.
View return policy
Description
May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.Specifications
| GLI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08151 | |
| GLI1 | |
| Human recombit protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B1 | |
| Unconjugated | |
| GLI1 | |
| GLI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2735 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction