Learn More
GLI1 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803812S
Encompass_Preferred
 View more versions of this product
Product Code. 17593073
  • € 242.00 / 30µL
Description
GLI1 Monoclonal antibody specifically detects GLI1 in Human samples. It is validated for Western Blot
Specifications
Specifications
GLI1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P08151
Mouse
Affinity chromatography
RUO
2735
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1F1
Unconjugated
GLI1
AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1
Human recombinant protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli.