missing translation for 'onlineSavingsMsg'
Learn More

GLI1 Monoclonal Antibody (OTI4B3), TrueMAB™, OriGene

Product Code. p-200015218
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17583173 30 μL 30µL
17563173 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17583173 Supplier Origene Technologies Supplier No. TA803888S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLI1 Monoclonal antibody specifically detects GLI1 in Human samples. It is validated for Western Blot

Specifications

Antigen GLI1
Applications Western Blot
Classification Monoclonal
Clone OTI4B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GLI1
Gene Accession No. P08151
Gene Alias AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2735
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.