missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144671
This item is not returnable.
View return policy
Description
Positive control: A549, LOVO, SH-SY5Y, human kidney tissue, human testis tissue. Predicted band size: 118 kDa Subcellular Location: Cytoplasm, Nucleus.
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| GLI1 | |
| Polyclonal | |
| Unconjugated | |
| GLI1 | |
| AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P08151 | |
| GLI1 | |
| Synthetic peptide corresponding to N terminal of Human Gli-1 aa 18264/1106. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction