missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572967
This item is not returnable.
View return policy
Description
GLI1 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| GLI1 | |
| Polyclonal | |
| Unconjugated | |
| GLI1 | |
| AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14632, 2735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol and 0.05% sodium azide | |
| P08151, P47806 | |
| GLI1 | |
| A synthetic peptide made to an internal portion of the human Gli1 protein (between residues 150-200). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction