Learn More
GLI2 Monoclonal Antibody (OTI1F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804549S
This item is not returnable.
View return policy
Description
GLI2 Monoclonal antibody specifically detects GLI2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| GLI2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96DB2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F9 | |
| Unconjugated | |
| HDAC11 | |
| AW546128; CJS; GLI family zinc finger 2; GLI family zinc finger protein 2; GLI2; GLI-Kruppel family member GLI2; glioma-associated oncogene family zinc finger 2; HPE9; oncogene GLI2; PHS2; Tax helper protein; tax helper protein 1; tax helper protein 2; tax-responsive element-2 holding protein; tax-responsive element-25-bp sequence binding protein; Thp; THP1; THP2; zinc finger protein GLI2 | |
| Full length human recombinant protein of human HDAC11 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |