Learn More
GLI2 Mouse anti-Human, Clone: OTI1G2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804599
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Gli-2 belongs to the C2H2-type zinc finger protein subclass of the Gli family. Members of this subclass are characterized as transcription factors which bind DNA through zinc finger motifs. These motifs contain conserved H-C links. Gli family zinc finger proteins are mediators of Sonic hedgehog (Shh) signaling and they are implicated as potent oncogenes in the embryonal carcinoma cell. The protein encoded by this gene localizes to the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. The encoded protein is associated with several phenotypes- Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, postaxial polydactyly types A1 and B.Specifications
| GLI2 | |
| Monoclonal | |
| Unconjugated | |
| GLI2 | |
| CJS, HPE9, PHS2, THP1, THP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2736 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1G2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P10070 | |
| GLI2 | |
| Human recombit protein fragment corresponding to amino acids 729-1123 of human GLI2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.