Learn More
GLI2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28245-1-AP-20UL
 View more versions of this product
Product Code. 18006675
  • € 145.00 / 20µL
Description
The GLI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GLI2. This antibody recognizes human antigen. The GLI2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GLI2
400 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
GLI family zinc finger 2, GLI2, HPE9, Tax helper protein, THP, THP1, THP2, Zinc finger protein GLI2
20 μL
2736
Store at -20°C. Stable for one year after shipment.