missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLI3 Recombinant Rabbit Monoclonal Antibody (K01_1R68)

Product Code. 30825023
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825023 50 μL
30825581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825023 Supplier Invitrogen™ Supplier No. MA565243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

GLI3 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1R68
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene GLI3
Gene Accession No. P10071
Gene Alias ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV_18031191mg; xgli3; xGLI-3; Xt; Zinc finger protein GLI3; zinc finger protein GLI3; transcriptional activator GLI3; zinc finger transcription factor Gli3
Gene Symbols GLI3
Host Species Rabbit
Immunogen Recombinant protein of Gli3.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2737
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.