missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI4 Antibody (OTI3A9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70833C
This item is not returnable.
View return policy
Description
GLI4 Monoclonal specifically detects GLI4 in Human samples. It is validated for Immunohistochemistry.
Specifications
| GLI4 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLI4 | |
| Full length recombinant protein of human GLI4 (NP_612474) produced in E.coli. | |
| 0.1 mL | |
| 2738.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI3A9 | |
| Immunohistochemistry | |
| GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction