missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI4 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806258S
This item is not returnable.
View return policy
Description
GLI4 Monoclonal antibody specifically detects GLI4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GLI4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30279 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 894 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI3A9 | |
| Unconjugated | |
| CCND2 | |
| 9130023H24Rik; GLI family zinc finger 4; GLI4; GLI-Kruppel family member GLI4; GLI-Kruppel family member GLI4 (oncogene HKR4); glioma-associated oncogene family zinc finger 4; HKR4; Krueppel-related zinc finger protein 4; Protein HKR4; RIKEN cDNA 9130023H24 gene; zinc finger protein GLI4; ZNF928 | |
| Full length human recombinant protein of human CCND2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction