missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gliquidone, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Gliquidone |
| CAS | 33342-05-1 |
| Analyte or Component Names | Gliquidone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H33 N3 O6 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenesulfonamide, N-[(cyclohexylamino)carbonyl]-4-[2-(3,4-dihydro-7-methoxy-4,4-dimethyl-1,3-dioxo-2(1H)-isoquinolinyl)ethyl]-, AR-DF 26, ARDF 26SE, Beglynor, Gliquidone, Glurenorm |
| InChI Formula | InChI=1S/C27H33N3O6S/c1-27(2)23-14-11-20(36-3)17-22(23)24(31)30(25(27)32)16-15-18-9-12-21(13-10-18)37(34,35)29-26(33)28-19-7-5-4-6-8-19/h9-14,17,19H,4-8,15-16H2,1-3H3,(H2,28,29,33) |
| Solution Type | Neat |
| SMILES | COc1ccc2c(c1)C(=O)N(CCc3ccc(cc3)S(=O)(=O)NC(=O)NC4CCCCC4)C(=O)C2(C)C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction