missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLMN Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97214PCP
This item is not returnable.
View return policy
Description
GLMN Polyclonal antibody specifically detects GLMN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GLMN | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FAP, FAP48FAP68, FKBPAP, GLMLFK506-binding protein-associated protein, glomulin, glomulin, FKBP associated protein, GVM, GVMFKBP-associated protein, VMGLOMvenous malformation with glomus cells | |
| Produced in rabbits immunized with E. coli-derived Human GLMN fragment. | |
| 0.1 mL | |
| Angiogenesis | |
| 11146 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction