missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLO1 Monoclonal Antibody (GT552)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531469
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (92%), Rat (91%), Xenopus laevis (82%), Rhesus Monkey (97%), Bovine (90%). Positive Control: 293T, HepG2, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
Specifications
| GLO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q04760, Q6P7Q4, Q9CPU0 | |
| GLO1 | |
| Recombinant protein encompassing a sequence within the center region of human Glyoxalase I. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT552 | |
| Unconjugated | |
| GLO1 | |
| 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035 | |
| Mouse | |
| Protein G | |
| RUO | |
| 109801, 2739, 294320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction