Learn More
Globe Scientific Dispenser For Borosilicate Glass Culture Tubes; for 100mm Tubes
Product Code. 31922585 Shop All Globe Scientific Products
Change view
Click to view available options
Capacity (Metric):
For 13 x 100mm tubes
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31922585 For 13 x 100mm tubes Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31922585 Supplier Globe Scientific Supplier No. 1592
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Metal Dispenser, for 13 x 100mm tubes
Metal dispenser
Heavy Duty dispenser
  • Made from white painted metal and feature a flexi-glass cover that pulls up to dispense the tubes
  • Each dispenser holds one box of 250 glass tubes
Specifications
Certifications/Compliance ISO 9001:2008, ASTM E 890, Type 1
Capacity (Metric) For 13 x 100mm tubes
Material Metal