Learn More
Novus Biologicals™ GLOD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29098
Product Code. 30047884
  • € 571.00 / 100µg
Description
GLOD4 Polyclonal antibody specifically detects GLOD4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLOD4
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
Unconjugated
C17orf25, CGI-150, chromosome 17 open reading frame 25, glyoxalase domain containing 4, glyoxalase domain-containing protein 4, HC71
Between 100 and 150
100 μg
Cancer
51031
Store at 4°C. Do not freeze.