missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-1R Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97308CL1
This item is not returnable.
View return policy
Description
GLP-1R Polyclonal antibody specifically detects GLP-1R in Canine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Dot Blot,Simple Western,KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| GLP-1R | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Dot Blot, Simple Western | |
| CoraFluor 1 | |
| GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220] | |
| 0.1 mL | |
| Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
| 2740 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction