missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-1R Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97308MFV500
This item is not returnable.
View return policy
Description
GLP-1R Polyclonal antibody specifically detects GLP-1R in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, KnockDown
Specifications
| GLP-1R | |
| Polyclonal | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated | |
| GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220] | |
| 0.1 mL | |
| Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
| 2740 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction