missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-2R Antibody (413801), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4285L
This item is not returnable.
View return policy
Description
GLP-2R Monoclonal antibody specifically detects GLP-2R in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| GLP-2R | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 413801 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| GLP-2 receptor, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor | |
| HEK293 human embryonic kidney cell line transfected with human GLP-2R, Met1-Ile553, Accession # O95838 | |
| 0.1 mL | |
| Cell Cycle and Replication, GPCR | |
| 9340 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction