missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-2R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | GLP-2R |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18497211
|
Novus Biologicals
NBP2-33557-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18112515
|
Novus Biologicals
NBP2-33557 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLP-2R Polyclonal specifically detects GLP-2R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GLP-2R | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O95838 | |
| 9340 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LPGVHELPMGIPAPWGTSPLSFHRKCSLWAPGRPFLTLVLLVSIKQVTGSLLEETTRKWAQYKQACLRDLLKEPSGIFCNGTFDQYVCWPHSSPGNVSVPC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GLP-2 receptor, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor | |
| GLP2R | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title