missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GLP/EHMT1 Antibody (1B7B4), Novus Biologicals™

Product Code. 30048451
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30048451 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30048451 Supplier Novus Biologicals™ Supplier No. NBP327913

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLP/EHMT1 Monoclonal antibody specifically detects GLP/EHMT1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP/EHMT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B7B4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10
Formulation PBS
Gene Accession No. Q9H9B1
Gene Alias EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 1H3-K9-HMTase 5, Eu-HMTase1DEL9q34, EUHMTASE1FP13812, FLJ12879, G9a-like protein 1, GLP, GLP1, H3 lysine-9 specific 5, KIAA1876euchromatic histone methyltransferase 1, KMT1DDKFZp667M072
Host Species Mouse
Immunogen Truncated recombinant GLP/EHMT1 expressed in E. Coli strain BL21 (DE3).
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 79813
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.