missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP1 Antibody (4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23558B
This item is not returnable.
View return policy
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human, Mouse, Mammalia samples. It is validated for ELISA, Immunohistochemistry
Specifications
| GLP1 | |
| Monoclonal | |
| Biotin | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| Synthetic GLP-1(7-17) | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 2641 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| 4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Mammalia | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction