missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLP1R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597790
This item is not returnable.
View return policy
Description
GLP1R Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Glucagon-like peptide-1(9-36)amide and GLP-1(9-37) are the forms of GLP-1 that result from the rapid degradation of the active forms of the peptide (GLP-1(7-36)amide and GLP-1(7-37)) by the enzyme dipeptidyl peptidase-IV (DPP-IV, also known as CD26 or adenosine deaminase binding protein). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.
Specifications
| GLP1R | |
| Polyclonal | |
| Unconjugated | |
| Glp1r | |
| Glip; GLP-1; GLP1 receptor; GLP-1 receptor; GLP1R; GLP-1R; GLP-1-R; GLP1Rc; GLP-I receptor; Glpr; glucagon like peptide 1 receptor; glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; LOW QUALITY PROTEIN: glucagon-like peptide 1 receptor; glucagon-like peptide 1 receptor; pancreatic beta cell receptor for the gluco-incretin hormone glucagon-like peptide 1; RATGL1RCP; seven transmembrane helix receptor | |
| Rabbit | |
| Protein G | |
| RUO | |
| 14652, 2740 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O35659, P43220 | |
| Glp1r | |
| Recombinant Human Glucagon-like peptide 1 receptor protein (24-145AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction