missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP2R, Mouse anti-Human, Clone: 1F2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009340-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: LEETTRKWAQYKQACLRDLLKEPSGIFCNGTFDQYVCWPHSSPGNVSVPCPSYLPWWSEESSGRAYRHCLAQGTWQTIENATDIWQDDSECSENHSFKQNVDRYALLSTLSpecifications
| GLP2R | |
| Monoclonal | |
| Unconjugated | |
| glucagon-like peptide 2 receptor | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1F2 | |
| In 1x PBS, pH 7.4 | |
| NM_004246 | |
| GLP2R | |
| GLP2R (NP_004237, 69 a.a. ∼ 178 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9340 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction