Learn More
Invitrogen™ GLRA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14661
Description
This antibody is specific for the ~48 kDa alpha 1 and alpha 2 subunits of the Glycine Receptor in Western blots of rat spinal cord and brain stem and in cell extracts, and does not recognize other Glycine Receptor subunits. Immunolabeling is blocked by preadsorption of antibody with the peptide immunogen. Reconstitute with 100 μL PBS.
GLRA1 is a subunit of a pentameric inhibitory glycine receptor, which mediates postsynaptic inhibition in the central nervous system. Defects in GLRA1 are a cause of startle disease (STHE), also known as hereditary hyperekplexia or congenital stiff-person syndrome. Multiple transcript variants encoding different isoforms have been found for GLRA1. Glycine receptors (GlyR) that include GLRA1 are members of the cys-loop family of ligand-gated ion channels, responsible for mediating the inhibitory effects of glycine. GLRA1 are widely distributed throughout the CNS, particularly within the hippocampus, spinal cord and brain stem. Diseases associated with GLRA1 include Hyperekplexia, Hereditary 1 and Hyperekplexia.
Specifications
| GLRA1 | |
| Polyclonal | |
| Unconjugated | |
| GLRA1 | |
| GLRA; Glra1; Glra2; glycine receptor; Glycine receptor 48 kDa subunit; glycine receptor alpha 1; glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-1; glycine receptor, alpha 1; glycine receptor, alpha 1 subunit; Glyr; GLYRA1; gra1; HKPX1; nmf11; oscillator; ot; spasmodic; spd; STHE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14654, 25674, 2741 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| 4 mg/mL | |
| 5mM Ammonium bicarbonate with no preservative; pH 7.4 | |
| P07727, P23415, Q64018 | |
| GLRA1 | |
| Synthetic peptide corresponding to the N-terminal region of rat GLRA1 conjugated to KLH. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |