missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS2 Monoclonal Antibody (OTI6D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809506
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a mitochondrial phosphate-activated glutaminase that catalyzes the hydrolysis of glutamine to stoichiometric amounts of glutamate and ammonia. This protein is functionally similar to the kidney glutaminase but is a little smaller in size. Originally thought to be liver-specific, this protein has been found in other tissues as well. At least one transcribed pseudogene has been found for this gene.Specifications
| GLS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UI32 | |
| GLS2 | |
| Full length human recombit protein of human GLS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI6D11 | |
| Unconjugated | |
| GLS2 | |
| GA, GLS, LGA, hLGA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27165 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction