missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTP Antibody (OTI2A10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71910AF488
This item is not returnable.
View return policy
Description
GLTP Monoclonal specifically detects GLTP in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GLTP | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLTP | |
| Full length recombinant protein of human GLTP (NP_057517) produced in HEK293T cells. | |
| 0.1 mL | |
| 51228.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2A10 | |
| Western Blot | |
| glycolipid transfer protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?