missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLTSCR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563267
This item is not returnable.
View return policy
Description
Immunogen sequence: ASSLDADEDG PMPSRNRPPI KTYEARSRIG LKLKIKQEAG LSKVVHNTAL DPVHQPPPPP ATLKVA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Component of SWI/SNF chromatin remodeling subcomplex GBAF that carries out key enzymatic activities, changing chromatin structure by altering DNA-histone contacts within a nucleosome in an ATP-dependent manner (PubMed:29374058). May play a role in BRD4-mediated gene transcription (PubMed:21555454). [UniProt].
Specifications
| GLTSCR1 | |
| Polyclonal | |
| Unconjugated | |
| BICRA | |
| BICRA; BRD4 interacting chromatin remodeling complex associated protein; BRD4 interacting chromatin remodelling complex associated protein; BRD4-interacting chromatin-remodeling complex-associated protein; glioma tumor suppressor candidate region gene 1; glioma tumor suppressor candidate region gene 1 protein; glioma tumor suppressor candidate region gene 1 protein; LOW QUALITY PROTEIN: BRD4-interacting chromatin-remodeling complex-associated protein; GLTSCR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29998 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZM4 | |
| BICRA | |
| Recombinant protein corresponding to Human GLTSCR1. Recombinant protein control fragment (Product #RP-98336). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction