missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Glucagon (1-29), bovine, human, porcine

Product Code. 16424987
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424987 Supplier MedChemExpress Supplier No. HYP0082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glucagon (1-29), bovine, human, porcine is a peptide hormone, produced by pancreatic α-cells. Glucagon stimulates gluconeogenesis. Glucagon (1-29), bovine, human, porcine activates HNF4α and increases HNF4α phosphorylation.

Specifications

Purity 95.13%
CAS 16941-32-5
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 3482.75 g/mol
Product Type Reference compound
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : 6.67 mg/mL (1.92 mM∣ultrasonic and adjust pH to 3 with HCl) ∣DMSO : 2.5 mg/mL (0.72 mM∣Need ultrasonic)
Synonym Porcine glucagon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.