missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon Antibody (181402), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1249T
This item is not returnable.
View return policy
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Glucagon | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 181402 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
| Human Glucagon synthetic peptide | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction, Stem Cell Markers | |
| 2641 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction