missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon Antibody (53), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05184JF549
This item is not returnable.
View return policy
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Mammalia samples. It is validated for Western Blot, ELISA
Specifications
| Glucagon | |
| Monoclonal | |
| Janelia Fluor 549 | |
| glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
| Synthetic GLP-1(9-18) | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction, Stem Cell Markers | |
| 2641 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 53 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mammalia | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction