missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Glucagon (Human, Porcine)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Glucagon (Human, Porcine) is an endogenous glucagon receptor agonist; regulates blood glucose.
Specifications
Specifications
| CAS | 16941-32-5 |
| Content And Storage | Store at -20°C |
| Description | Endogenous glucagon receptor agonist |
| Formulation | C153H225N43O49S |
| Molecular Weight (g/mol) | 3482.78 |
| Quantity | 1 mg |
| Sequence | HSQGTFTSDYSKYLDSRRAQDFVQWLMNT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction