missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon Mouse anti-Human, Rat, Porcine, Clone: 9, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
€ 210.00 - € 500.00
Specifications
| Antigen | Glucagon |
|---|---|
| Clone | 9 |
| Concentration | 1 mg/ml |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:8000, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin 1:8000, Sandwich ELISA 1:100-1:2000, Sandwich ELISA Capture |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18431442
|
Novus Biologicals
NBP2-21803 |
0.1 ml |
€ 500.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132623
|
Novus Biologicals
NBP2-21803-0.025ML |
0.025 mL |
€ 222.00 € 210.00 / 25µL Save € 12.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucagon Monoclonal specifically detects Glucagon in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Sandwich ELISA Capture.Specifications
| Glucagon | |
| 1 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 2641 | |
| IgG1 κ | |
| Protein A or G purified |
| 9 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:8000, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin 1:8000, Sandwich ELISA 1:100-1:2000, Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| Cancer, GPCR, Signal Transduction, Stem Cell Markers | |
| glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
| Synthetic C-terminal fragment of glucagon coupled to carrier. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title