missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glucagon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589937
This item is not returnable.
View return policy
Description
Positive Samples: BxPC-3, 293T, Rat spleen Immunogen sequence: RSLQDTEEKS RSFSASQADP LSDPDQMNED KRHSQGTFTS DYSKYLDSRR AQDFVQWLMN TKRNRNNIA.
Glucagon is a 29-residue polypeptide hormone (MW 3482), produced in the pancreas. A related hormone, enteroglucagon (or oxyntomodulin), which is produced in the mucosa of the small and large intestine, consists of the 29 amino acid sequence of pancreatic glucagon extended by 8 additional residues at the C-terminus. The biological activities of pancreatic glucagon include glycogenolysis, lipolysis, gluconeogenesis, and ketogensis, which are antagonistic effects to those of insulin action, thus leading to increased blood glucose levels. Immunocytochemical studies have revealed the presence of pancreatic glucagon inside the A or alpha cells, which constitute 15-20% of the islet cell population. These cells are located preferentially at the periphery of the human pancreatic islets. Pathological manifestations of the glucagon-type peptide residue almost exclusively with the exsistence of tumors or glucagonomas, as no states of glucagon-cell deficiency or hyperplasia have been identified. Glucagon-specific antibodies would prove useful as a cell and tumor markers applying immunohistochemical techniques, and as an analytical tool in qualification of the hormone.
Specifications
| Glucagon | |
| Polyclonal | |
| Unconjugated | |
| Gcg | |
| GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; IL12p35; IL12P40; Incretin hormone; OXM; OXY; Oxyntomodulin; porcine proglucagon; PPG; preproglucagon; proglucagon | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14526, 24952, 2641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.82 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01275, P06883, P55095 | |
| Gcg | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction