Learn More
Glucagon R/GCGR Antibody (591929) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10296H
This item is not returnable.
View return policy
Description
Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Glucagon R/GCGR | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 591929 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
| HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871 | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2642 | |
| Store at 4°C in the dark. | |
| IgG2a |