missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon R/GCGR Antibody (crotedumab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28852JF635
This item is not returnable.
View return policy
Description
Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Glucagon R/GCGR | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
| GCGR | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2642 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| crotedumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction