missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glucagon Receptor Recombinant Human Monoclonal Antibody (9G1)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA543157
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Glucagon receptors are G-protein coupled receptors for glucagon and play a central role in the regulation of blood glucose levels and glucose homeostatsis. It regulates the rate of hepatic glucose production by promoting glycogen hydrolysis and gluconeogenesis. Glucagon receptors also play an important role in mediating the responses to fasting. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G-proteins) and modulates the activity of down-stream effectors.
Specifications
| Glucagon Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P47871 | |
| GCGR | |
| Recombinant Human GCGR protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 9G1 | |
| Unconjugated | |
| GCGR | |
| GCGR; GGR; GL-R; Glucagon receptor; glucagon receptor perhaps same as Niddm3; GR | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 2642 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction