missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucokinase/GCK Antibody (SR1917), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22233-50ul
This item is not returnable.
View return policy
Description
Glucokinase/GCK Monoclonal antibody specifically detects Glucokinase/GCK in Human samples. It is validated for Western Blot
Specifications
| Glucokinase/GCK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1917 | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
| A synthesized peptide derived from human Glucokinase/GCK (Uniprot #: P35557) | |
| 50 μg | |
| Lipid and Metabolism, Protein Kinase | |
| 2645 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction