missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 433.00 - € 572.00
Specifications
| Antigen | Glucosaminyl (N-acetyl) Transferase 1/GCNT1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18461001
|
Novus Biologicals
NBP1-86809-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18209536
|
Novus Biologicals
NBP1-86809 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 Polyclonal specifically detects Glucosaminyl (N-acetyl) Transferase 1/GCNT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Glucosaminyl (N-acetyl) Transferase 1/GCNT1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2650 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LINLCGMDFPIKTNLEIVRKLKLLMGENNLETERMPSHKEERWKKRYEVVNGKLTNTGTVKMLPPLETPLFSGSAYFVVSREYVGYVLQNEKIQKLMEWAQDTYSPDEYLWA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| beta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N-acetylglucosaminyltransferase, beta-1,6-N-acetylglucosaminyltransferase, C2GNT1, C2GNTbeta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N--acetylglucosaminyltransferase, C2GNT-L, core 2 beta1,6 N-acetylglucosaminyltransferase-I, core 2 beta-1,6-N-acetylglucosaminyltransferase I, core 2 branching enzyme, Core 2 GNT, Core 2-branching enzyme, Core2-GlcNAc-transferase, EC 2.4.1.102, G6NT, glucosaminyl (N-acetyl) transferase 1, core 2, glucosaminyl (N-acetyl) transferase 1, core 2(beta-1,6-N-acetylglucosaminyltransferase), MGC126335, MGC126336, NACGT2, NAGCT2 | |
| GCNT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title