missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 1-dehydrogenase Antibody (OTI2A7), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71478C
This item is not returnable.
View return policy
Description
Glucose 1-dehydrogenase Monoclonal specifically detects Glucose 1-dehydrogenase in Human, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Glucose 1-dehydrogenase | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| H6PD | |
| Full length recombinant protein of human Glucose 1-dehydrogenase (NP_004276) produced in HEK293T cells. | |
| 0.1 mL | |
| 9563.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2A7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 6-phosphogluconolactonase, DKFZp686A01246, EC 1.1.1.49, EC 2.7.4.3, EC 3.1.1.31, G6PD, H form, G6PDH, GDH/6PGL endoplasmic bifunctional protein, GDHglucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary, hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase), MGC87643 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction