missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose Standard for YSI2300 Glucose/Lactate Analyzer, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
120 mL
Unit Size:
Each
Description
- Glucose Standard for YSI2300 Glucose/Lactate Analyzer.
Specifications
Specifications
| Packaging | HDPE Bottle |
| Physical Form | Liquid |
| For Use With (Equipment) | YSI2300 Glucose/Lactate Analyzer |
| Quantity | 120 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction