missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glucose Transporter GLUT8 Antibody (VB1-050) - Humanized, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glucose Transporter GLUT8 Monoclonal antibody specifically detects Glucose Transporter GLUT8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Glucose Transporter GLUT8 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | VB1-050 |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | Q9NY64 |
| Gene Alias | GLUT-8, GLUT8solute carrier family 2 (facilitated glucose transporter) member 8, GLUTX1facilitated glucose transporter member 8, solute carrier family 2 (facilitated glucose transporter), member 8 |
| Host Species | Human |
| Immunogen | SLC2A8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?