missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114036
This item is not returnable.
View return policy
Description
Immunogen sequence: MYRYLAKALL PSRAGPAALG SAANHSAALL GRGRGQPAAA SQPGLALAAR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
Glutamate dehydrogenase 2, Glud2, is localized to the mitochondrion and acts as a homohexamer to recycle glutamate during neurotransmission. The enzyme catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate. Diseases associated with GLUD2 include Parkinson Disease, Late-Onset and Hyperinsulinemic Hypoglycemia, Familial, 6. Among its related pathways are GABA shunt and Carbon metabolism.
Specifications
| GluD2 | |
| Polyclonal | |
| Unconjugated | |
| GLUD2 | |
| B230104L07Rik; cpr; creeper; GDH 2; GDH2; GluD2; GLUDP1; GluR delta2; gluR delta-2 subunit; GLURD2; GluRdelta2; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; glutamate ionotropic receptor delta type subunit 2; glutamate receptor delta-2 subunit; glutamate receptor ionotropic, delta-2; glutamate receptor, ionotropic, delta 2; Grid2; ho; hotfoot; Lc; Lc>J>; lurcher; minisatellite 10ac detected by probe MMS10; MMS10-AC; Ms10ac; nmf408; SCAR18; testicular secretory protein Li 14; tpr | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2747 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P49448 | |
| GLUD2 | |
| Synthetic peptide directed towards the N terminal region of human GLUD2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction