missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1, p Ser845 Antibody (SR1027), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21554-50ul
This item is not returnable.
View return policy
Description
GluR1 Monoclonal antibody specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| GluR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1027 | |
| Western Blot 1:500-1:1000 | |
| AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
| A synthesized peptide derived from human phospho-GluR1 [Ser845] (Uniprot #: P42261) | |
| 50 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
| 2890 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction