missing translation for 'onlineSavingsMsg'
Learn More

GluR2 Antibody (3A11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-29731B

Product Code. 18482062

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GluR2 Monoclonal antibody specifically detects GluR2 in Human, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GluR2
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Rat, Primate
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
3A11
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
AMPA 2, glutamate receptor, ionotropic, AMPA 2
Recombinant fusion protein TrpE-GluR2.
0.1 mL
Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
2891
Store at 4C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.