missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2 Antibody (3A11), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29731JF646
This item is not returnable.
View return policy
Description
GluR2 Monoclonal antibody specifically detects GluR2 in Human, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| GluR2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Primate | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| 3A11 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| Recombinant fusion protein TrpE-GluR2. | |
| 0.1 mL | |
| Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
| 2891 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction