missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluR7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598452
This item is not returnable.
View return policy
Description
GluR7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics.
Specifications
| GluR7 | |
| Polyclonal | |
| Unconjugated | |
| GRIK3 | |
| 9630027E11; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GLUR7; Glur-7; GluR7-3; GluR7a; glutamate ionotropic receptor kainate type subunit 3; glutamate receptor; glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 3; Grik3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2899 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13003 | |
| GRIK3 | |
| Recombinant Human Glutamate receptor ionotropic, kainate 3 protein (32-240AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction