missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody (GLUT1/2476) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75787JF635
This item is not returnable.
View return policy
Description
Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Glut1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 6513 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| GLUT1/2476 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction