Learn More
Invitrogen™ GLUT1 Monoclonal Antibody (10C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549148
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human SLC2A1 shares 98% and 98.3% amino acid (aa) sequence identity with mouse and rat SLC2A1, respectively. Positive Control - WB: human placenta tissue, human placenta tissue. IHC: human placenta tissue, human renal cancer tissue. ICC/IF: SiHa cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Glucose transporter 1 (GLUT1), encoded by the SLC2A1 gene, is a facilitative glucose transporter that plays a pivotal role in the uptake of glucose across cell membranes, particularly in tissues with high-energy demands such as the brain, erythrocytes, and endothelial cells of the blood-brain barrier. GLUT1 is ubiquitously expressed and functions through a passive diffusion mechanism, allowing glucose to move down its concentration gradient. Its involvement in maintaining glucose homeostasis is crucial for cellular metabolism and energy production. Mutations in SLC2A1 can lead to GLUT1 deficiency syndrome, characterized by seizures, developmental delay, and movement disorders due to impaired glucose transport into the brain. GLUT1 also plays a significant role in cancer biology, where its overexpression is commonly observed in various tumors, facilitating the enhanced glucose uptake required for rapid tumor cell proliferation. This characteristic makes GLUT1 a potential target for cancer diagnostics and therapeutics. Additionally, GLUT1's regulation is influenced by factors such as hypoxia, insulin, and cellular stress, highlighting its adaptive role in different physiological and pathological states.
Specifications
| GLUT1 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P11166 | |
| SLC2A1 | |
| E.coli-derived human SLC2A1 recombinant protein (Position: R92-V492). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10C10 | |
| Unconjugated | |
| SLC2A1 | |
| choreoathetosis/spasticity, episodic (paroxysmal choreoathetosis/spasticity); CSE; DYT17; DYT18; DYT9; EIG12; glucose transporter glut1; glucose transporter type 1, erythrocyte/brain; glucose transporter type I; GLUT; Glut1; Glut-1; GLUT1DS; GLUTB; GT1; GTG1; Gtg3; HepG2 glucose transporter; HTLVR; human T-cell leukemia virus (I and II) receptor; MGC141895; MGC141896; PED; RATGTG1; receptor for HTLV-1 and HTLV-2; SDCHCN; Slc2a1; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2 a 1 (facilitated glucose transporter) brain; solute carrier family 2 member 1; solute carrier family 2, facilitated glucose transporter member 1; solute carrier family 2, member 1 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 6513 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.