Learn More
GLUT1 Mouse anti-Human, Unconjugated, Clone: 2A5A2, Proteintech
Product Code. 16888025 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16888025 150 μL 150µL
16898025 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16888025 Supplier Proteintech Supplier No. 662901IG150UL
Mouse Monoclonal Antibody
The GLUT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GLUT1. This antibody recognizes human, mouse antigen. The GLUT1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Antigen GLUT1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 2A5A2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 6513
Target Species Human, Mouse
Isotype IgG1